missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CapG Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00258CL1
This item is not returnable.
View return policy
Description
CapG Polyclonal antibody specifically detects CapG in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| CapG | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| actin-regulatory protein CAP-G, AFCP, capping protein (actin filament), gelsolin-like, gelsolin-like capping protein, macrophage capping protein, macrophage-capping protein, MCPActin regulatory protein CAP-G | |
| Produced in rabbits immunized with purified, recombinant Human CapG (Accession#: AAH14549.1; Met1-Lys348) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 822 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction