missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAPG Monoclonal Antibody (K52030_2E1), Invitrogen™

Mouse Monoclonal Antibody

€ 480.00 - € 846.00

Specifications

Antigen CAPG
Clone K52030_2E1
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30207493
View Documents
Invitrogen™
MA555844
50 μL
€ 480.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30202415
View Documents
Invitrogen™
MA556224
200 μL
€ 846.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: RDLALAIRDS ERQGKAQVEI VTDGEEPAEM IQVLGPKPAL KEGNPEEDLT ADKANAQAAA LYKVSDATGQ MNLTKVADSS

CAPG Monoclonal antibody specifically detects CAPG in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

CAPG
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Actin regulatory protein CAP-G; actin-capping protein GCAP39; actin-regulatory protein CAP-G; AFCP; Capg; capping actin protein, gelsolin like; capping protein (actin filament), gelsolin-like; epididymis secretory protein Li 66; gCap39; gelsolin-like capping protein; HEL-S-66; macrophage capping protein; macrophage-capping protein; mbh1; MCP; myc basic motif homolog 1
Recombinant human CAPG (immunogen range is 191-270).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
CAPG
K52030_2E1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
P40121
822
IgG1
Protein A
Antibody
Videos
SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado