Learn More
Invitrogen™ CAPG Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5104443
Product Code. 16640895
  • € 502.00 / 100µL
Description
Antibody detects endogenous levels of total CAPG.
This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in non-muscle cells. Alternatively spliced transcript variants have been observed, but have not been fully described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CAPG
Polyclonal
Unconjugated
CAPG
Actin regulatory protein CAP-G; actin-capping protein GCAP39; actin-regulatory protein CAP-G; AFCP; Capg; capping actin protein, gelsolin like; capping protein (actin filament), gelsolin-like; epididymis secretory protein Li 66; gCap39; gelsolin-like capping protein; HEL-S-66; macrophage capping protein; macrophage-capping protein; mbh1; MCP; myc basic motif homolog 1
Rabbit
Affinity Chromatography
RUO
12332, 297339, 822
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P24452, P40121, Q6AYC4
CAPG
A synthesized peptide derived from human CAPG(Accession P40121), corresponding to amino acid residues L170-M220.
100 μL
Primary
Human, Mouse, Rat