missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19535-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in non-muscle cells. Alternatively spliced transcript variants have been observed for this gene.Specifications
| CAPG | |
| Polyclonal | |
| Unconjugated | |
| CAPG | |
| Actin regulatory protein CAP G, AFCP, CAPG, Macrophage capping protein, MCP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12332, 297339, 822 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24452, P40121, Q6AYC4 | |
| CAPG | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction