missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capicua Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-59906F
This item is not returnable.
View return policy
Description
Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| Capicua | |
| Polyclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
| Rabbit | |
| 250 kDa | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 23152 | |
| Store at 4C in the dark. |
| Immunocytochemistry, Immunofluorescence, KnockDown | |
| 1mg/mL | |
| Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| Q96RK0 | |
| CIC | |
| The epitope recognized by this antibody maps to a C-terminal region within residue 1500-1608 of human Capicua. [NP_055940.3] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction