missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capicua Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-59905MFV610
This item is not returnable.
View return policy
Description
Capicua Polyclonal antibody specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Western Blot (Negative)
Specifications
| Capicua | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Western Blot (Negative) | |
| capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
| The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3] | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 23152 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry, Immunoprecipitation | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction