Learn More
Trajan Scientific and Medical™ Capillary Electrophoresis Tubing
Click to view available options
Diameter (English) Inner:
0.001 in.
0.002 in.
0.003 in.
0.004 in.
Unit Size:
1m
Description
- Manufactured from precision bore tubing
- Wide pH range
- 30, 50, 75 and 100μm ID
- Column-to-column reproducibility
- Untreated capillary electrophoresis tubing
Specifications
Specifications
| Type | Electrophoresis Tubing Untreated 0.363mm OD x 0.075mm ID x 1m Length |
| Description | Untreated |
| Finish | Hydrophobic and hydrophilic surface treatments |
| Diameter (English) Inner | 0.003 in. |
| Diameter (Metric) Inner | 0.075 mm |
| Diameter (Metric) Outer | 0.363 mm |
| Overall Length (Metric) | 1 m |
| Material | Precision Bore Tubing |