missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPN2 Mouse anti-Canine, Human, Clone: OTI3G1, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504280
This item is not returnable.
View return policy
Description
Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.Specifications
| CAPN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17655 | |
| CAPN2 | |
| Full length human recombit protein of human CAPN2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G1 | |
| Unconjugated | |
| CAPN2 | |
| CANP2, CANPL2, CANPml, mCANP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 480118, 824 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction