Learn More
CAPN2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1E1F10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66977-100UL
Description
The CAPN2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAPN2. This antibody recognizes human, mouse, rat antigen. The CAPN2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| CAPN2 | |
| 1E1F10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| Calpain 2 catalytic subunit, Calpain 2 large subunit, Calpain large polypeptide L2, Calpain M type, CANP 2, CANP2, CANPL2, CANPml, CAPN2, FLJ39928, M calpain, mCANP, Millimolar calpain | |
| 100 μL | |
| 824 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |