Learn More
CAPN2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1E1F10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66977-20UL
 View more versions of this product
Product Code. 18010326
  • € 145.00 / 20µL
Description
The CAPN2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAPN2. This antibody recognizes human, mouse, rat antigen. The CAPN2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
CAPN2
1E1F10
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
Calpain 2 catalytic subunit, Calpain 2 large subunit, Calpain large polypeptide L2, Calpain M type, CANP 2, CANP2, CANPL2, CANPml, CAPN2, FLJ39928, M calpain, mCANP, Millimolar calpain
20 μL
824
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.