Learn More
CAPS, 0.5M buffer soln., pH 9.0, Thermo Scientific™
Product Code. 15475929
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475929 250 mL 250mL
15485929 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15475929 Supplier Thermo Scientific Alfa Aesar Supplier No. J63749.AK
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1135-40-6
Concentration 0.5 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Molecular Formula C9H19NO3S
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
SMILES OS(=O)(=O)CCCNC1CCCCC1
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
Molecular Weight (g/mol) 221.32