missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPSL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561337
This item is not returnable.
View return policy
Description
Immunogen sequence: GIKGLGRVFR IMDDDNNRTL DFKEFMKGLN DYAVVMEKEE VEELFRRFDK DG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
CAPSL gene ontology annotations related to this gene include calcium ion binding.
Specifications
| CAPSL | |
| Polyclonal | |
| Unconjugated | |
| Capsl | |
| 1700028N11Rik; AW125205; Calcyphosine; calcyphosine like; calcyphosine-like; calcyphosin-like protein; CAPSL; RGD1308776 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 133690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWF8 | |
| Capsl | |
| Recombinant protein corresponding to Human CAPSL. Recombinant protein control fragment (Product #RP-100435). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction