missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPSL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17174-1-AP-150UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CAPSL gene ontology annotations related to this gene include calcium ion binding.Tekniske data
| CAPSL | |
| Polyclonal | |
| Unconjugated | |
| CAPSL | |
| Calcyphosin like protein, calcyphosine like, CAPSL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 133690, 294795, 75568 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P8Y1, Q8WWF8 | |
| Capsl | |
| CAPSL Fusion Protein Ag10771 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion