missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPSL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17174-1-AP-20UL
This item is not returnable.
View return policy
Description
CAPSL gene ontology annotations related to this gene include calcium ion binding.Specifications
| CAPSL | |
| Polyclonal | |
| Unconjugated | |
| CAPSL | |
| Calcyphosin like protein, calcyphosine like, CAPSL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 133690, 294795, 75568 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P8Y1, Q8WWF8 | |
| Capsl | |
| CAPSL Fusion Protein Ag10771 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction