Learn More
Thermo Scientific™ CaptureSelect™ hGH Ligand Leakage ELISA
Alle ansehen Thermo Scientific ProdukteBeschreibung
CaptureSelect hGH Ligand Leakage ELISA Kit is designed to detect possible leached ligand of human growth hormone (hGH) purified using the CaptureSelect hGH affinity matrix. The hGH affinity ligand, a 13 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The hGH Ligand Leakage ELISA Kit can be used for optimal purification process development and in routine quality control of in-process streams as well as final product.
Spezifikation
Spezifikation
| Content And Storage | Store in freezer (-5°C to -30°C). |
| Assay | ELISA |
| For Use With (Equipment) | Microplate Reader |
| Shipping Condition | Dry Ice |
| Product Line | CaptureSelect |
| Quantity | 1 Assay |
| Type | ELISA Kit |
| Format | Kit |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.