missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZA1 Antibody (OTI2G4), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70336PE
This item is not returnable.
View return policy
Description
CAPZA1 Monoclonal specifically detects CAPZA1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| CAPZA1 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| CAPZA1 | |
| Full length recombinant protein of human CAPZA1 (NP_006126) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI2G4 | |
| Flow Cytometry | |
| Cap Z, CAPPA1, capping protein (actin filament) muscle Z-line, alpha 1, CAPZ, CapZ alpha-1, CAZ1, F-actin-capping protein subunit alpha-1 | |
| Mouse | |
| Affinity Purified | |
| Extracellular Matrix | |
| 829.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction