missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbaryl-d7 (naphthyl-d7), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Carbaryl-d7 |
| CAS | 362049-56-7 |
| Molecular Formula | C12 2H7 H4 N O2 |
| Quantity | 0.05 g |
| Synonym | 1-Naphthalen-2,3,4,5,6,7,8-d7-ol, methylcarbamate (9 CI), 1-Naphthalenyl-2,3,4,5,6,7,8-d7 N-methylcarbamate |
| InChI Formula | InChI=1 S/C12H11NO2/c1-13-12(14)15-11-8-4-6-9-5-2-3-7-10(9)11/h2-8 H,1H3,(H,13,14)/i2D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(OC(=O)NC)c([2 H])c([2 H])c([2 H])c2c1[2 H] |
| IUPAC Name | (2,3,4,5,6,7,8-heptadeuterionaphthalen-1-yl) N-methylcarbamate |
| Molecular Weight (g/mol) | 208.26 |
| Formula Weight | 208.1229 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction