missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbohydrate Sulfotransferase 10/CHST10 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97238AF750
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 10/CHST10 Polyclonal antibody specifically detects Carbohydrate Sulfotransferase 10/CHST10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Carbohydrate Sulfotransferase 10/CHST10 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| carbohydrate sulfotransferase 10, EC 2.8.2, HNK-1 sulfotransferase, HNK-1STEC 2.8.2.-, HNK1STHuHNK-1ST, huHNK-1ST, MGC17148 | |
| Produced in rabbits immunized with E. coli-derived Human Carbohydrate Sulfotransferase 10/CHST10 fragment. | |
| 0.1 mL | |
| Cancer, CD Markers, Endocrinology, Immunology, Signal Transduction | |
| 9486 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction