Learn More
Carbolite™ Sample Tray for AAF 11/3
Product Code. 11926149
Change view
Click to view available options
For Use With (Equipment):
AAF 11/3 Ashing Furnace
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11926149 AAF 11/3 Ashing Furnace Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11926149 Supplier Carbolite™ Supplier No. 0016732051
  • ONLINE EXCLUSIVE PRICE
    € 213.14
    € 101.89 / Pack of 1
    Save € 111.25
    52% Off
View all promotions
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable sample tray for the powerful and versatile AAF Ashing Furnace. Carbolite™ Sample Tray, AAF 11/3 should be used with a loading handle.
  • Suitable for AAF 11/3 Ashing Furnace
  • Use with a loading handle for safety
  • Durable and heat-resistant
Specifications
For Use With (Equipment) AAF 11/3 Ashing Furnace
Type Sample Tray