Learn More
Corning™ Carbolite Furnace
Product Code. 10228127 Shop All Corning Products
Change view
Click to view available options
:
Size: 254 x 356mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10228127 Size: 254 x 356mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10228127 Supplier Corning™ Supplier No. 3286
Specifications
For Use With (Equipment) Growing anchorage dependent cells
Includes an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings
No. per Case 1
Product Type Culture System Kit
Culture Area 8500 cm²
Quantity 1Pack