Learn More
Thermo Scientific™ Carbonate-buffered saline (5X), pH 9.0
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Spécification
| Concentration | 5X |
| pH Range | 9.0 |
| Format | Liquid |
| Quantity | 500 mL |
| Storage Requirements | Ambient temperatures |
| Type | Carbonate-buffered saline |