missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase I/CA1 Antibody (26E3), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Carbonic Anhydrase I/CA1 |
|---|---|
| Clone | 26E3 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30037213
|
Novus Biologicals
NBP3-26469-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028628
|
Novus Biologicals
NBP3-26469-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic Anhydrase I/CA1 Monoclonal antibody specifically detects Carbonic Anhydrase I/CA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
| Carbonic Anhydrase I/CA1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CA-I, Car1, Carbonate dehydratase I, carbonic anhydrase 1, Carbonic anhydrase B, carbonic anhydrase ICAB, carbonic dehydratase, EC 4.2.1.1 | |
| A synthesized peptide derived from Human Carbonic Anhydrase I/CA1 [UniProt P00915] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 26E3 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Hypoxia, Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 759 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title