missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase I/CA1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-6618-0.2ml
This item is not returnable.
View return policy
Description
Carbonic Anhydrase I/CA1 Polyclonal antibody specifically detects Carbonic Anhydrase I/CA1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Carbonic Anhydrase I/CA1 | |
| Polyclonal | |
| Western Blot 1:2000-1:10000, ELISA 1:20000-1:100000 | |
| P00915, P00915, P00915, P00915 | |
| Goat | |
| Delipidation and Defibrination | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| CA-I, Car1, Carbonate dehydratase I, carbonic anhydrase 1, Carbonic anhydrase B, carbonic anhydrase ICAB, carbonic dehydratase, EC 4.2.1.1 | |
| Carbonic Anhydrase I/CA1/CA1 [Human Erythrocytes] (Uniprot: P00915) | |
| 0.2 mL | |
| Hypoxia, Lipid and Metabolism | |
| 759 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction