missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase II/CA2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 224.00 - € 481.00
Specifications
| Antigen | Carbonic Anhydrase II/CA2 |
|---|---|
| Applications | Western Blot, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18381097
|
Novus Biologicals
NBP3-03387-20UL |
20 μL |
€ 224.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18322216
|
Novus Biologicals
NBP3-03387-100UL |
100 μL |
€ 481.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic Anhydrase II/CA2 Polyclonal specifically detects Carbonic Anhydrase II/CA2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
| Carbonic Anhydrase II/CA2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human Carbonic Anhydrase II/CA2 (NP_000058.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Hypoxia, Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 760 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title