Learn More
Invitrogen™ Carbonic Anhydrase II Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578897
Beskrivelse
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human THP-1 whole cell, human HEL whole cell, rat kidney tissue, rat stomach tissue, mouse kidney tissue, mouse stomach tissue. IHC: human gastric cancer tissue, human liver cancer tissue. Flow: HT-29 cell, SW480 cell.
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
Tekniske data
| Carbonic Anhydrase II | |
| Polyclonal | |
| Unconjugated | |
| CA2 | |
| AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12349, 54231, 760 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P00918, P00920, P27139 | |
| CA2, Car2 | |
| E.coli-derived human CA2 recombinant protein (Position: S2-K260). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.