Learn More
Invitrogen™ Carbonic Anhydrase II Recombinant Rabbit Monoclonal Antibody (23GB3300), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553401
 View more versions of this product
Product Code. 30089058
  • € 207.00 / 20µL
Description
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carbonic Anhydrase II
Recombinant Monoclonal
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P00918
CA2
A synthetic peptide of human Carbonic anhydrase 2 (200-260AA).
20 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB3300
Unconjugated
CA2
AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5
Rabbit
Affinity chromatography
RUO
760
-20°C