missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Carbonic Anhydrase III/CA3 Antibody (CA3/7884), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26745-20ug
This item is not returnable.
View return policy
Description
Carbonic Anhydrase III/CA3 Monoclonal antibody specifically detects Carbonic Anhydrase III/CA3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Carbonic Anhydrase III/CA3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| CAIII, CA-III, Car3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, EC 4.2.1.1 | |
| Recombinant fragment of human Carbonic Anhydrase III/CA3 protein (exact sequence is proprietary) | |
| 20 μg | |
| Lipid and Metabolism | |
| 761 | |
| Store at 4°C. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| CA3/7884 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction