missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody (09), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26103G
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IX/CA9 Monoclonal antibody specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Carbonic Anhydrase IX/CA9 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 09 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Carbonic Anhydrase IX/CA9 (Accession#: NP_001207.2, Met1-Asp414) | |
| 0.1 mL | |
| Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers | |
| 768 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction