missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody (SPM314), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34408CL1
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IX/CA9 Monoclonal antibody specifically detects Carbonic Anhydrase IX/CA9 in Equine,Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Carbonic Anhydrase IX/CA9 | |
| Monoclonal | |
| CoraFluor 1 | |
| CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| Microsomal fraction of human renal cortical tissue homogenate | |
| 0.1 mL | |
| Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers | |
| 768 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF, Immunocytochemistry/Immunofluorescence | |
| SPM314 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Equine, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction