missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VB/CA5B Antibody (030), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90091FR
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VB/CA5B Monoclonal specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for ELISA.
Specifications
| Carbonic Anhydrase VB/CA5B | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CA2 Human CA3 Human CA4 Human CA5A Human CA8 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 30 | |
| ELISA | |
| Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CA-VB, EC 4.2.1.1, MGC39962 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase VB/CA5B (Q9Y2D0; Cys34-Pro 317). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 11238 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction