Learn More
Carbonic Anhydrase VB/CA5B Antibody (030), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90091MFV610
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VB/CA5B Monoclonal antibody specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for ELISA
Specifications
| ELISA | |
| 11238 | |
| IgG |
| Primary | |
| Store at 4C in the dark. |