missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VI Antibody (401809), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB29391CL1
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VI Monoclonal antibody specifically detects Carbonic Anhydrase VI in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Carbonic Anhydrase VI | |
| Monoclonal | |
| CoraFluor 1 | |
| Carbonate dehydratase VI, carbonic anhydrase 6, carbonic anhydrase VIGUSTIN, CA-VI, EC 4.2.1.1, MGC21256, Salivary carbonic anhydrase, Secreted carbonic anhydrase | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase VI, Gln18-Asn308, Accession # EAW71606 | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 765 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 401809 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction