missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VIII/CA8 Antibody (005), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90710CL1
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VIII/CA8 Monoclonal antibody specifically detects Carbonic Anhydrase VIII/CA8 in Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Carbonic Anhydrase VIII/CA8 | |
| Monoclonal | |
| CoraFluor 1 | |
| CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase VIII/CA8 (Uniprot#: P28651; Met1-Gln291). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 767 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 005 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction