missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Carbonic Anhydrase VIII/CA8 Antibody (rCA8/8835), Novus Biologicals™
Mouse Monoclonal Antibody
€ 232.00 - € 462.00
Specifications
| Antigen | Carbonic Anhydrase VIII/CA8 |
|---|---|
| Clone | rCA8/8835 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043991
|
Novus Biologicals™
NBP3-26748-20ug |
20 μg |
€ 232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043306
|
Novus Biologicals™
NBP3-26748-100ug |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic Anhydrase VIII/CA8 Monoclonal antibody specifically detects Carbonic Anhydrase VIII/CA8 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Carbonic Anhydrase VIII/CA8 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 767 | |
| IgG1 κ | |
| Protein A or G purified |
| rCA8/8835 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 | |
| Recombinant full-length human Carbonic Anhydrase VIII/CA8 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title