missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase VIII Recombinant Rabbit Monoclonal Antibody (001)

Product Code. 16353784
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16353784 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16353784 Supplier Invitrogen™ Supplier No. MA530212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse Carbonic Anhydrase VIII/CA8.

The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase VIII
Applications ELISA
Classification Recombinant Monoclonal
Clone 001
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Car8
Gene Accession No. P28651
Gene Alias AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118
Gene Symbols Car8
Host Species Rabbit
Immunogen Recombinant Mouse Carbonic Anhydrase VIII/CA8 protein (Met1-Gln291).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12319
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.