missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase X/CA10 Antibody (7H8F12E6), Alexa Fluor™ 405 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06193AF405
This item is not returnable.
View return policy
Description
Carbonic Anhydrase X/CA10 Monoclonal antibody specifically detects Carbonic Anhydrase X/CA10 in Human samples. It is validated for ELISA
Specifications
| Carbonic Anhydrase X/CA10 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| carbonic anhydrase X, carbonic anhydrase-related protein 10, Carbonic anhydrase-related protein X, CARP X, CA-RPX, CARPXCA-RP X, Cerebral protein 15, cerebral protein-15, HUCEP-15 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Carbonic Anhydrase X/CA10 (NP_001076002.1; Met 1-Asn 300). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 7H8F12E6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 56934 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction