missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XII/CA12 Antibody (08), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26096B
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XII/CA12 Monoclonal antibody specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for ELISA
Specifications
| Carbonic Anhydrase XII/CA12 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 08 | |
| ELISA | |
| Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
| This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 771 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction