missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XIII/CA13 Antibody (103), FITC , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89511F
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XIII/CA13 Monoclonal antibody specifically detects Carbonic Anhydrase XIII/CA13 in Human samples. It is validated for ELISA
Specifications
| Carbonic Anhydrase XIII/CA13 | |
| Monoclonal | |
| FITC | |
| carbonic anhydrase 13, carbonic anhydrase XIIICarbonate dehydratase XIII, CAXIII, CA-XIII, EC 4.2.1.1, FLJ37995, MGC59868 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase XIII/CA13 (NP_940986.1; Met1-His262). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 377677 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 103 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction