missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XIII/CA13 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99317JF646
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XIII/CA13 Polyclonal antibody specifically detects Carbonic Anhydrase XIII/CA13 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| carbonic anhydrase 13, carbonic anhydrase XIIICarbonate dehydratase XIII, CAXIII, CA-XIII, EC 4.2.1.1, FLJ37995, MGC59868 | |
| Antigen and protein A Affinity-purified | |
| Lipid and Metabolism | |
| Store at 4C in the dark. |
| Rabbit | |
| 0.1 mL | |
| Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction