missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XIV/CA14 Antibody (004), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89508FR
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XIV/CA14 Monoclonal specifically detects Carbonic Anhydrase XIV/CA14 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Carbonic Anhydrase XIV/CA14 | |
| Monoclonal | |
| DyLight 680 | |
| Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase XIV/CA14 extracellular domain (NP_036245.1; Met 1-Met 290). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 23632 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CAIX/ CA9 Human CA10/ CARPX Human CA12/ CAXII Mouse CA14/ Car14 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction