missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XIV/CA14 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98207B
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XIV/CA14 Polyclonal specifically detects Carbonic Anhydrase XIV/CA14 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Carbonic Anhydrase XIV/CA14 | |
| Polyclonal | |
| Western Blot, ELISA | |
| Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
| Produced in rabbits immunized with purified, recombinant Carbonic Anhydrase XIV/CA14 (NP_035927.1; Met 1-Met 290). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 23632 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Biotin | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction