missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XIV/CA14 Rabbit anti-Human, Clone: 110, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 552.00
Specifications
| Antigen | Carbonic Anhydrase XIV/CA14 |
|---|---|
| Clone | 110 |
| Dilution | Sandwich ELISA Capture 1:250-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18237127
|
Novus Biologicals
NBP2-89509-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18209027
|
Novus Biologicals
NBP2-89509-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic Anhydrase XIV/CA14 Monoclonal specifically detects Carbonic Anhydrase XIV/CA14 in Human samples. It is validated for Sandwich ELISA Capture.Specifications
| Carbonic Anhydrase XIV/CA14 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
| Recombinant Human Carbonic Anhydrase XIV/CA14 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 110 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2 um filtered solution in PBS | |
| 23632 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title