missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carboxypeptidase A1 Monoclonal Antibody (CL6629)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531441
This item is not returnable.
View return policy
Description
Immunogen sequence: DFVGHQVLRI SVADEAQVQK VKELEDLEHL QLDFWRGPAH PGSPIDVRVP FPSIQAVKIF LESHGISYET Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 77%.
CPA1, also known as CPA, is a 419 amino acid secreted monomeric protein that is highly expressed in pancreatic tissue. Functioning as a pancreatic exopeptidase, CPA1 uses zinc as a cofactor to catalyze the release of C-terminal amino acids from a variety of proteins, thereby playing a key role in protein digestion and degradation. Via its catalytic activity, CPA1 is also thought to be involved in zymogen (proenzyme) inhibition, probably functioning to block enzyme activation pathways. Abnormal levels of CPA1 are associated with pancreatic cancer, suggesting a possible role in either tumor progression or tumor suppression events.
Specifications
| Carboxypeptidase A1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P15085 | |
| Cpa1 | |
| Recombinant protein fragment of Human CPA1 (Product #RP-92630) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL6629 | |
| Unconjugated | |
| Cpa1 | |
| 0910001L12Rik; carboxypeptidase A; carboxypeptidase A1; carboxypeptidase A1 (pancreatic); carboxypeptidase A1, pancreatic; CPA; Cpa1; pancreatic carboxypeptidase A; pancreatic procarboxypeptidase A | |
| Mouse | |
| Protein A | |
| RUO | |
| 1357 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction