missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A2/CPA2 Antibody (009), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89530MFV500
This item is not returnable.
View return policy
Description
Carboxypeptidase A2/CPA2 Monoclonal antibody specifically detects Carboxypeptidase A2/CPA2 in Human samples. It is validated for ELISA
Specifications
| Carboxypeptidase A2/CPA2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1358 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 009 | |
| Sandwich ELISA Detection | |
| carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase A2/CPA2 (Accession#: NP_001859.1; Met1-Tyr417). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction