Learn More
Carboxypeptidase A2/CPA2 Antibody (133), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90716APC
Product Code. 18645411
  • € 607.00 / 0.10mL
Description
Carboxypeptidase A2/CPA2 Monoclonal antibody specifically detects Carboxypeptidase A2/CPA2 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1 mL