missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carboxypeptidase A2 Monoclonal Antibody (2E11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518635
This item is not returnable.
View return policy
Description
Peptide Sequence: NFGAYHTLEE ISQEMDNLVA EHPGLVSKVN IGSSFENRPM NVLKFSTGGD KPAIWLDAGI HAREWVTQAT ALWTANKIVS DYGKDPSIT*.
Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.
Specifications
| Carboxypeptidase A2 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P48052 | |
| CPA2 | |
| CPA2 (NP_001860, 117 a.a. ~ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 2E11 | |
| Unconjugated | |
| CPA2 | |
| carboxypeptidase A2; carboxypeptidase A2 (pancreatic); carboxypeptidase A2, pancreatic; CBPA2; Cpa2; EC 3.4.17; EC 3.4.17.15; LOW QUALITY PROTEIN: carboxypeptidase A2; zgc:92530 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1358 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction