missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase B1/CPB1 Antibody (329), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89533MFV500
This item is not returnable.
View return policy
Description
Carboxypeptidase B1/CPB1 Monoclonal antibody specifically detects Carboxypeptidase B1/CPB1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Carboxypeptidase B1/CPB1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1360 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 329 | |
| ELISA, Immunohistochemistry-Paraffin | |
| carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase B1/CPB1 (Accession#: NP_001862.2; Met 1-Tyr 417). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction