missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase B2/CPB2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98368AF532
This item is not returnable.
View return policy
Description
Carboxypeptidase B2/CPB2 Polyclonal specifically detects Carboxypeptidase B2/CPB2 in Mouse samples. It is validated for ELISA.
Specifications
| Carboxypeptidase B2/CPB2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Alexa Fluor 532 | |
| carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor | |
| Produced in rabbits immunized with purified, recombinant Carboxypeptidase B2/CPB2 (Q9JHH6; Met1-Thr422). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1361 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction