missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase D Polyclonal Antibody
Carboxypeptidase D Polyclonal Antibody
Brand: Invitrogen PA575825
This item is not returnable.
View return policy
Description
Carboxypeptidase D Polyclonal Antibody for Western Blot
Specifications
| Carboxypeptidase D | |
| Polyclonal | |
| Unconjugated | |
| CPD | |
| AA960140; AW322530; carboxypeptidase D; CPD; cpd {ECO:0000312; D830034L15Rik; gp180; metallocarboxypeptidase D; RGD:2393} | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1362 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75976 | |
| CPD | |
| Synthetic peptide corresponding to residues in Human Carboxypeptidase D. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction