missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase E/CPE Antibody (1), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89258CL1
This item is not returnable.
View return policy
Description
Carboxypeptidase E/CPE Monoclonal antibody specifically detects Carboxypeptidase E/CPE in Human samples. It is validated for ELISA
Specifications
| Carboxypeptidase E/CPE | |
| Monoclonal | |
| CoraFluor 1 | |
| carboxypeptidase E, Carboxypeptidase H, CPH, EC 3.4.17, EC 3.4.17.10, Enkephalin convertase, Prohormone-processing carboxypeptidase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase E/CPE (Accession#: NP_001864.1; Met 1-Ser 453). | |
| 0.1 mL | |
| Neuroscience | |
| 1363 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 1 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction